Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-fluoro-4-chromanone hydantoin

Catalog No. CDS004715 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004715 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004715 Supplier Sigma Aldrich Supplier No. CDS004715100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isopropyl butyrate is an electroantennographic detection (ead) active compound that was identified by GC-mass spectrometry
  • It is attractive to both sexes of west indian fruit fly, anastrepha obliqua (macquart) (diptera: tephritidae).isopropyl butyrate was used as intravaginal taggant in the study of breath test for vaginal gel use

Specifications

Molecular Formula C11H9FN2O3
Linear Formula C11H9FN2O3
MDL Number MFCD01569247
Quantity 100 mL
Molecular Weight (g/mol) 236.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.