Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-o-anisaldehyde 98.0+%, TCI America™
SDP

Catalog No. F11755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11755G 5 g
F11751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11755G Supplier TCI America Supplier No. F11755G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146137-74-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD01090998
InChI Key UIOAYOIJMYMOEU-UHFFFAOYSA-N
Synonym 2-methoxy-6-fluorobenzaldehyde, 6-fluoro-o-anisaldehyde, 2-fluoro-6-methoxy-benzaldehyde, 2-fluoro-6-mehoxybenzaldehyde, benzaldehyde, 2-fluoro-6-methoxy, 2-fluoro-6-methoxybenzaldehyde, pubchem8486, acmc-209yud, 3-fluoro-2-formylanisole, intermediates-zcf02117
PubChem CID 2737357
IUPAC Name 2-fluoro-6-methoxybenzaldehyde
SMILES COC1=C(C=O)C(F)=CC=C1

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-o-anisaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.