Learn More
Medchemexpress LLC 6-Fluoroindole | 399-51-9 | 135.14 | 1 ML
SDP
Catalog No. 5000449053
Encompass_Preferred
6-Fluoroindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethylene-indoles, which are potential anticancer immunomodulators.
  • Employed as an antifungal and antibacterial agent.
  • Serves as a potent selective serotonin reuptake inhibitor.
  • Used as an inhibitor of HIV-1 attachment.
Catalog No. 50-004-49053 Supplier Medchemexpress LLC Supplier No. HYW00227110MM/1ML
6-Fluoroindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethylene-indoles, which are potential anticancer immunomodulators.
  • Employed as an antifungal and antibacterial agent.
  • Serves as a potent selective serotonin reuptake inhibitor.
  • Used as an inhibitor of HIV-1 attachment.