Learn More
Medchemexpress LLC 6-Fluoroindole | 399-51-9 | 135.14 | 25 G
SDP
Catalog No. 5000449436
Encompass_Preferred
6-Fluoroindole is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It is intended for research use only and not for patient use.
  • Reagent in the synthesis of tryptophan dioxygenase inhibitors (pyridyl-ethylene-indoles), which are potential anticancer immunomodulators
  • Antifungal and antibacterial agent
  • Potent selective serotonin reuptake inhibitor
  • Inhibitor of HIV-1 attachment
Catalog No. 50-004-49436 Supplier Medchemexpress LLC Supplier No. 25GHYW002271
6-Fluoroindole is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It is intended for research use only and not for patient use.
  • Reagent in the synthesis of tryptophan dioxygenase inhibitors (pyridyl-ethylene-indoles), which are potential anticancer immunomodulators
  • Antifungal and antibacterial agent
  • Potent selective serotonin reuptake inhibitor
  • Inhibitor of HIV-1 attachment