Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluorosalicylic acid, 98+%, Thermo Scientific™

Catalog No. AC398770010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC398770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398770010 Supplier Thermo Scientific Chemicals Supplier No. 398770010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 67531-86-6
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
InChI Key BCEKGWWLVKXZKK-UHFFFAOYSA-N
Synonym 6-fluorosalicylic acid, 6-fluoro-2-hydroxybenzoic acid, 2-fluoro-6-hydroxybenzoicacid, 6-fluorosalicylicacid, benzoic acid, 2-fluoro-6-hydroxy, 2-fluoro-6-hydoxybenzoic acid, pubchem4958, acmc-1baav, 2-carboxy-3-fluorophenol
PubChem CID 586777
IUPAC Name 2-fluoro-6-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)O

Specifications

Melting Point 162°C to 164°C
Assay Percent Range 98% min. (HPLC)
Packaging Glass Bottle
Linear Formula FC6H3(OH)COOH
Quantity 1 g
Formula Weight 156.11
Percent Purity 98+%
Chemical Name or Material 6-Fluorosalicylic acid
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.