Learn More
Sigma Aldrich 6-Fluorotryptamine hydrochloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- This product is informer compound x12 of the aryl halide chemistry informer library developed by chemists at merck & co., inc., kenilworth, nj, u.s.., inc., kenilworth, nj, u.s., which contains 18 drug-like molecules representative of those encountered in complex synthesis
- By screening a new reaction against the informer library, chemists can directly compare and analyze a reactions's successes and shortcomings among different methods and various research teams
- It may also be used to faciliate deeper method development for performance or utility.1.chemistry informer libraries: a chemoinformatics enabled approach to evaluate and advance synthetic methods, chem
- Sci., 2016, 7, 2604-26132.nanomole-scale high-throughput chemistry for the synthesis of complex molecules
Specifications
Specifications
| CAS | 55206-24-1 |
| Melting Point | 233°C to 235°C (lit.) |
| Molecular Formula | C10H11FN2 -+ HCl |
| Linear Formula | C10H11FN2 -+ HCl |
| MDL Number | MFCD00012687 |
| Quantity | 100 mg |
| Synonym | 3-(2-Aminoethyl)-6-fluoroindole hydrochloride |
| Molecular Weight (g/mol) | 214.67 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.