Learn More
Abcam 6-FMOC-Amino-D-luciferin, 10MG
Supplier:  Abcam AB27591910MG
Encompass_Preferred
MW 501.59 g/mol, Purity >98%. Amino analog of D-luciferin substrate for firefly luciferase activity. Can be used in automated peptide synthesis for incorporation of amino-luciferin into synthetic peptides.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2739