Learn More
Research Products International Corp 6"" Foam Insert, 60x1.5-2.0ml
SDP
Catalog No. 501978178
Encompass
Two high-density foam inserts to accommodate up to 60 microtubes. This insert fits into the 6 Inch (152mm) Platform attachment for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Catalog No. 50-197-8178 Supplier Research Products International Corp Supplier No. 155308
Two high-density foam inserts to accommodate up to 60 microtubes. This insert fits into the 6 Inch (152mm) Platform attachment for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.