Learn More
Electron Microscopy Sciences 6" Forceps with Curved "B" Tips, Stainless Steel
SDP
Catalog No. 5029023
Encompass_Preferred
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Curved "B" Tips
Catalog No. 50-290-23 Supplier Electron Microscopy Sciences Supplier No. 729936C
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Curved "B" Tips