Learn More
Electron Microscopy Sciences 6" Forceps with Straight Tips, Stainless Steel
SDP
Catalog No. 5029020
Encompass_Preferred
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Straight Tips
Catalog No. 50-290-20 Supplier Electron Microscopy Sciences Supplier No. 729956S
Made from stainless steel, medium points, serrated tips and alignment pin, curved or straight tips. Only curved type A tips comes with locking device.
Tips: Straight Tips