Learn More
Enzo Life Sciences 6-Formylindolo(3,2-b)carbazole, (100µg), CAS Number: 172922-91-7
SDP
Supplier:  Enzo Life Sciences BMLGR2060100
Encompass_Preferred
Ah receptor ligand. A tryptophan-derived, high affinity aryl hydrocarbon receptor ligand (Kd=7x10-11 M). Proposed to be an endogenous AhR ligand. Induces rapid and transient expression of cytochrome P450-1A1 (CYP1A1) in human cells at 100 pM. Alternative Name: FICZ. Formula: C19H12N2O. MW: 284.3. CAS Number: 172922-91-7. Purity: ≥95% (HPLC). Appearance: Yellow solid. Solubility: Soluble in DMSO (10mg/ml). Use/Stability: Store solutions at -20°C for up to one month. Long Term Storage: -20°C
Catalog No. 50-103-1779