Learn More
Cayman Chemical 6-FormylpterIn 10mg
SDP
Catalog No. 502955871
Encompass_Preferred
An oxidized pterin that binds to one of two active sites on XO, resulting in “hetero-substrate” inhibition at nanomolar concentrations; converted by XO to 6-carboxylpterin and H2O2, acting as an intracellular generator of H2O2 in cells expressing XO
Catalog No. 50-295-5871 Supplier Cayman Chemical Supplier No. 1424710 MG
An oxidized pterin that binds to one of two active sites on XO, resulting in “hetero-substrate” inhibition at nanomolar concentrations; converted by XO to 6-carboxylpterin and H2O2, acting as an intracellular generator of H2O2 in cells expressing XO