Learn More
Princeton Cryotech 6 FT TRANSFER LN2 CRYO HOSE
Catalog No. NC0278273
Encompass_Preferred
  • Designed for efficient transfer of liquid nitrogen.
  • Constructed with a stainless steel spiral wrap casing.
  • Features a 1/2 in stainless steel female swivel flare nut (CGA 295) end connection.
  • Handles cryogenic applications safely.
  • Maximum allowable working pressure of 1200 PSI.
  • Standard cryogenic transfer hose for reliable performance.
  • Suitable for various LN2 system requirements.
  • 6 ft length for versatile use.
  • Inner diameter of 1/2 in for optimal flow.
Catalog No. NC0278273 Supplier Princeton Cryotech Supplier No. 9713109
  • Designed for efficient transfer of liquid nitrogen.
  • Constructed with a stainless steel spiral wrap casing.
  • Features a 1/2 in stainless steel female swivel flare nut (CGA 295) end connection.
  • Handles cryogenic applications safely.
  • Maximum allowable working pressure of 1200 PSI.
  • Standard cryogenic transfer hose for reliable performance.
  • Suitable for various LN2 system requirements.
  • 6 ft length for versatile use.
  • Inner diameter of 1/2 in for optimal flow.