Learn More
Medchemexpress LLC HY-14615 10mg , [6]-Gingerol CAS:23513-14-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1461510MG
Encompass_Preferred
HY-14615 10mg [6]-Gingerol CAS: 23513-14-6 [6]-Gingerol is an active compound isolated from Ginger (Zingiber officinale Rosc), exhibits a variety of biological activities including anticancer, anti-inflammation, and anti-oxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0133