Learn More
Medchemexpress LLC HY-14615 5mg Medchemexpress, [6]-Gingerol CAS:23513-14-6 Purity:>98%
SDP
Catalog No. 502081625
Encompass_Preferred
Medchemexpress, HY-14615 5mg [6]-Gingerol CAS:23513-14-6 [6]-Gingerol is an active compound isolated from Ginger (Zingiber officinale Rosc), exhibits a variety of biological activities including anticancer, anti-inflammation, and anti-oxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1625 Supplier Medchemexpress LLC Supplier No. HY146155MG
Medchemexpress, HY-14615 5mg [6]-Gingerol CAS:23513-14-6 [6]-Gingerol is an active compound isolated from Ginger (Zingiber officinale Rosc), exhibits a variety of biological activities including anticancer, anti-inflammation, and anti-oxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.