Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[6]-Gingerol, Analytical Standard, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018056 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8056 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8056 Supplier MilliporeSigma Supelco Supplier No. 5086610MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[6]-Gingerol is a naturally occurring plant phenoland an active pungent constituent found in the rhizome of ginger, which is known to possess anti-inflammatory, anti-tumor and antioxidant properties and can hence, serve as a potential candidate in the treatment of cancer.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 23513-14-6
Physical Form Neat
Linear Formula C17H26O4
MDL Number MFCD00210507
Synonym 3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5 S)-; 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone; 6-Gingerol
Quantity 10 mg
Formula Weight 294.39
Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.