Learn More
Medchemexpress LLC 6-Hepten-1-ol 5g
SDP
Catalog No. 5000206611
Encompass_Preferred
6-Hepten-1-ol has a floral and green odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes air fresheners and cleaning agents
Catalog No. 50-002-06611 Supplier Medchemexpress LLC Supplier No. HYW1274755G
6-Hepten-1-ol has a floral and green odor and is commonly used as a flavoring agent in the food and beverage industry In addition it can be used as an intermediate in the synthesis of various organic compounds including fragrances pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes notably in the production of perfumes air fresheners and cleaning agents