Learn More
Medchemexpress LLC 6-heptenoic acid, 2-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-2-methyl-, (2R)- | 288617-77-6 | MFCD09879889 | 98.8% | 379.45 | C23H25NO4 | 1 G
SDP
Catalog No. 5000301331
Encompass_Preferred
Fmoc-(R)-2-(pentenyl)Ala-OH is an Fmoc-protected, chiral alkenyl amino acid used as a building block in peptide synthesis. It provides an (R)-configured 2-(pentenyl) side chain that can be incorporated into peptides during solid-phase or solution-phase synthesis and subsequently modified by alkenyl-selective reactions such as ring-closing metathesis to produce stapled peptides or other constrained analogs.
  • Fmoc-protected for compatibility with standard Fmoc solid-phase peptide synthesis.
  • Chiral (R) configuration for stereospecific incorporation.
  • Alkenyl side chain suitable for ring-closing metathesis and other transformations.
  • Applicable to both solid-phase and solution-phase peptide assembly.
  • High purity to support consistent coupling and analytical characterization.
  • Available in laboratory-scale pack sizes for research applications.
Catalog No. 50-003-01331 Supplier Medchemexpress LLC Supplier No. HYW0188471G
Fmoc-(R)-2-(pentenyl)Ala-OH is an Fmoc-protected, chiral alkenyl amino acid used as a building block in peptide synthesis. It provides an (R)-configured 2-(pentenyl) side chain that can be incorporated into peptides during solid-phase or solution-phase synthesis and subsequently modified by alkenyl-selective reactions such as ring-closing metathesis to produce stapled peptides or other constrained analogs.
  • Fmoc-protected for compatibility with standard Fmoc solid-phase peptide synthesis.
  • Chiral (R) configuration for stereospecific incorporation.
  • Alkenyl side chain suitable for ring-closing metathesis and other transformations.
  • Applicable to both solid-phase and solution-phase peptide assembly.
  • High purity to support consistent coupling and analytical characterization.
  • Available in laboratory-scale pack sizes for research applications.