Learn More
Medchemexpress LLC 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, sodium salt (1:1), (3R,5S,6E)- | 147098-18-8 | 99.9% | 503.52 | 200 MG
SDP
Catalog No. 5000386888
Encompass_Preferred
Rosuvastatin Sodium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM. It potently blocks hERG current with an IC50 of 195 nM. This compound effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.
  • Reduces the expression of mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein.
  • Effectively lowers low-density lipoprotein (LDL) cholesterol.
  • Reduces triglyceride levels.
  • Decreases C-reactive protein levels.
Catalog No. 50-003-86888 Supplier Medchemexpress LLC Supplier No. HY17504B200MG
Rosuvastatin Sodium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM. It potently blocks hERG current with an IC50 of 195 nM. This compound effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.
  • Reduces the expression of mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein.
  • Effectively lowers low-density lipoprotein (LDL) cholesterol.
  • Reduces triglyceride levels.
  • Decreases C-reactive protein levels.