Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Heptynenitrile 98.0+%, TCI America™
SDP

Catalog No. H12145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12145G 5 g
H121425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12145G Supplier TCI America Supplier No. H12145G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15295-69-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
InChI Key BIGRRVLEKCQFHS-UHFFFAOYSA-N
Synonym 6-Cyano-1-hexyne
PubChem CID 11768526
IUPAC Name hept-6-ynenitrile
SMILES C#CCCCCC#N

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 3276
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Heptynenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.