Learn More
AdipoGen 6-HEX dipivaloate
SDP
Supplier:  AdipoGen CDXC0187M025
Encompass_Preferred
Chemical. CAS 1166837-63-3. Formula C31H22Cl6O9. MW 751.2274. Synthetic. 6-HEX is an amino-conjugatable fluorescent probe that is widely used in nucleic acid sequencing and related research. It can also be used to label peptides and oligonucleotides.
Catalog No. 50-256-1431