Learn More
Aat Bioquest 6-HEX, SE [6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein, succinimidyl ester] *Single Isomer*
SDP
Catalog No. 501952807
Encompass_Preferred
6-HEX, SE is the amine-reactive succinimidyl ester of 6-HEX acid. It is widely used in nucleic acid sequencing and related research.
Catalog No. 50-195-2807 Supplier Aat Bioquest Supplier No. 202
Add to Cart
Edge
Add to Cart
6-HEX, SE is the amine-reactive succinimidyl ester of 6-HEX acid. It is widely used in nucleic acid sequencing and related research.