Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-2-methylquinoline 98.0+%, TCI America™
SDP

Catalog No. H14461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14461G 1 g
H14465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14461G Supplier TCI America Supplier No. H14461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-21-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00047600
InChI Key USSQQASIZNTRAJ-UHFFFAOYSA-N
Synonym 6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g
PubChem CID 594395
IUPAC Name 2-methylquinolin-6-ol
SMILES CC1=NC2=C(C=C1)C=C(C=C2)O

Specifications

Melting Point 216°C
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.