Learn More
Medchemexpress LLC HY-W001171 100mg Medchemexpress, 3-Hydroxyanthranilic acid CAS:548-93-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW001171100MG
Encompass_Preferred
Medchemexpress, HY-N6658 100mg 6-Hydroxy-4-methylcoumarin CAS:2373-31-1 6-Hydroxy-4-methylcoumarin (compound 9) is a coumarins secondary metabolites and has anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0635