Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. H10055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10055G Supplier TCI America Supplier No. H10055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2373-31-1
Melting Point 248°C
Molecular Formula C10H8O3
Quantity 5 g
MDL Number MFCD00016967
Synonym 6-hydroxy-4-methylcoumarin, 6-hydroxy-4-methyl-2h-chromen-2-one, coumarin, 6-hydroxy-4-methyl, 6-hydroxy-4-methyl-2-benzopyrone, 6-hydroxy-4-methyl-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy-4-methyl, chembl12626, 2h-1-benzopyran-2-one,6-hydroxy-4-methyl, acmc-1cuho, 4-methyl-6-hydroxycoumarin
InChI Key IRUHWRSITUYICV-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=C(C=C2)O
IUPAC Name 6-hydroxy-4-methylchromen-2-one
Molecular Weight (g/mol) 176.171
PubChem CID 75409
Formula Weight 176.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-4-methylcoumarin
Show More Show Less
RTECSNumber GN6999800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.