Learn More
Medchemexpress LLC 6-hydroxy-7-methoxycoumarin | 776-86-3 | 192.17 | 5 MG
SDP
Catalog No. 5000416363
Encompass_Preferred
Isoscopoletin is an active constituent found in Artemisia argyi leaves. It demonstrates inhibitory activity against HBV replication and exhibits anti-inflammatory effects by inhibiting the MAPK/NF-κB/STAT/AKT signaling pathway. It also significantly inhibits cell proliferation.
  • Significantly inhibits cell proliferation in human CCRF-CEM leukaemia cells and multidrug-resistant subline CEM/ADR5000 cells.
  • Demonstrates inhibitory activity against HBV replication.
  • Exhibits anti-inflammatory effects by inhibiting the MAPK/NF-κB/STAT/AKT signaling pathway.
  • Inhibits intracellular glucose consumption and lactate production in tumor cells.
  • Downregulates levels of TARC, MDC, MCP-1, IL-8, and IL-1β in HaCaT cells.
Catalog No. 50-004-16363 Supplier Medchemexpress LLC Supplier No. HYN13655MG
Add to Cart
Add to Cart
Isoscopoletin is an active constituent found in Artemisia argyi leaves. It demonstrates inhibitory activity against HBV replication and exhibits anti-inflammatory effects by inhibiting the MAPK/NF-κB/STAT/AKT signaling pathway. It also significantly inhibits cell proliferation.
  • Significantly inhibits cell proliferation in human CCRF-CEM leukaemia cells and multidrug-resistant subline CEM/ADR5000 cells.
  • Demonstrates inhibitory activity against HBV replication.
  • Exhibits anti-inflammatory effects by inhibiting the MAPK/NF-κB/STAT/AKT signaling pathway.
  • Inhibits intracellular glucose consumption and lactate production in tumor cells.
  • Downregulates levels of TARC, MDC, MCP-1, IL-8, and IL-1β in HaCaT cells.