Learn More
Medchemexpress LLC 6β-hydroxy cortisol-d4 | 1261254-51-6 | MFCD11656667 | 98.9% | 382.48 g/mol | C21H26D4O6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW587724S1MG
Encompass_Preferred
6β-Hydroxy Cortisol-d4 is a deuterium-labeled analogue of 6β-hydroxy cortisol supplied as a dry powder for use as a stable isotope internal standard and tracer in quantitative bioanalytical assays. It is intended for analytical workflows and compatible with common mass spectrometry and NMR techniques.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable as a tracer in NMR, GC-MS, and LC-MS methods.
  • High purity, typically about 98.9%.
  • Supplied as a dry powder for long-term storage at -20°C.
  • Provided in small research quantities (for example, 1 mg).
Catalog No. 50-002-85116