Learn More
Cayman Chemical 6-hydroxy Melatonin, Purity: ≥95%, CAS: 2208-41-5, 10 mg
SDP
Catalog No. 502985843
Encompass_Preferred
An active metabolite of melatonin; a MT1A, MT1B, and MT2 receptor agonist; inhibits dopamine release from isolated rabbit retina (IC50 = 0.0016 µM); reduces increases in the levels of NF-κB, IL-6, and IL-8 and decreases in the levels of GSH in LPS- and peptidoglycan G-stimulated HUVECs at 10 and 100 µM; reduces iron-induced lipid oxidation in rat hippocampal homogenate at 10 mg/kg
Catalog No. 50-298-5843 Supplier Cayman Chemical Supplier No. 2185710 MG
An active metabolite of melatonin; a MT1A, MT1B, and MT2 receptor agonist; inhibits dopamine release from isolated rabbit retina (IC50 = 0.0016 µM); reduces increases in the levels of NF-κB, IL-6, and IL-8 and decreases in the levels of GSH in LPS- and peptidoglycan G-stimulated HUVECs at 10 and 100 µM; reduces iron-induced lipid oxidation in rat hippocampal homogenate at 10 mg/kg