Learn More
Cayman Chemical 6-hydroxy WarfarIn 10mg
SDP
Catalog No. 502892945
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin
Catalog No. 50-289-2945 Supplier Cayman Chemical Supplier No. 900063210 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin