Learn More
Cayman Chemical 6-hydroxy Warfarin, Purity: ≥98%, CAS: 17834-02-5, 1 mg
SDP
Catalog No. 502892594
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin
Catalog No. 50-289-2594 Supplier Cayman Chemical Supplier No. 90006321 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin