Learn More
Cayman Chemical 6-hydroxy WarfarIn 500UG
SDP
Catalog No. 502893326
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin
Catalog No. 50-289-3326 Supplier Cayman Chemical Supplier No. 9000632500 UG
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin