Learn More
Cayman Chemical 6-hydroxy WarfarIn 5mg
SDP
Catalog No. 502893921
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin
Catalog No. 50-289-3921 Supplier Cayman Chemical Supplier No. 90006325 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9, CYP1A1, and CYP1A2 metabolism of (+)-warfarin