Learn More
AdipoGen 6-Hydroxybenzothiazole-2-CN
SDP
Catalog No. 502561395
Encompass_Preferred
Chemical. CAS 939-69-5. Formula C8H4N2OS. MW 176.2. Synthetic. Bulding block for synthesis. Key intermediate for the synthesis of luminescent reagent D-luciferin.
Catalog No. 50-256-1395 Supplier AdipoGen Supplier No. CDXC0263GG25
Chemical. CAS 939-69-5. Formula C8H4N2OS. MW 176.2. Synthetic. Bulding block for synthesis. Key intermediate for the synthesis of luminescent reagent D-luciferin.