Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxycaproic acid, 95%

Catalog No. AC342240250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC342240250 25 g Glass bottle
AC342240010 1 g Glass bottle
AC342240050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC342240250 Supplier Thermo Scientific Chemicals Supplier No. 342240250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1191-25-9 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-25-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00046560
InChI Key IWHLYPDWHHPVAA-UHFFFAOYSA-N
Synonym 6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
PubChem CID 14490
ChEBI CHEBI:17869
IUPAC Name 6-hydroxyhexanoic acid
SMILES C(CCC(=O)O)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 38.0°C to 40.0°C
Color White
Density 0.9810g/mL
Boiling Point 113.0°C to 116.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.44
Linear Formula OH(CH2)5COOH
Quantity 25 g
Specific Gravity 0.981
Formula Weight 132.16
Percent Purity 95%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material 6-Hydroxycaproic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.