Learn More
Medchemexpress LLC HY-N6656 100mg Medchemexpress, 6-Hydroxycoumarin CAS:6093-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN6656100MG
Encompass_Preferred
Medchemexpress, HY-N6656 100mg 6-Hydroxycoumarin CAS:6093-68-1 6-Hydroxycoumarin is a coumarin which has anti-inflammatory, anti-pyretic, anti-oxidant, vasodilator, anti-amoebic, anti-bacterial, anti-fungal, bacteriostatic and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2953