Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxycoumarin 98.0+%, TCI America™
SDP

Catalog No. H132725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H132725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H132725G Supplier TCI America Supplier No. H132725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6093-68-1
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00630040
InChI Key CJIJXIFQYOPWTF-UHFFFAOYSA-N
PubChem CID 99477
IUPAC Name 6-hydroxychromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1O

Specifications

Melting Point 255°C
Color Yellow
Quantity 25 g
Synonym 6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
Formula Weight 162.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.