Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-hydroxyflavanone | 4250-77-5 | MFCD00017485 | 99.9% | 240.25 g/mol | C15H12O3 | 1 G
SDP

Catalog No. 5000299334
Encompass_Preferred

6-Hydroxyflavanone is a naturally occurring flavanone used as a research chemical with reported activity against cyclooxygenase-2 (COX-2), 5-lipoxygenase (5-LOX), and opioid and GABA-A receptors; it is employed in studies of inflammation, neuropathic pain, and diabetes research.

  • High purity (99.87%) suitable for analytical and biological studies.
  • Chemical formula C15H12O3; molecular weight 240.25 g/mol.
  • Available as a powder for convenient storage and handling.
  • Targets COX-2, 5-LOX, opioid receptors, and GABA-A receptors in assays.
  • Reported applications include inflammation, neuropathic pain, and diabetes research.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term).
  • For research use only; not for human or veterinary applications.

Catalog No. 50-002-99334 Supplier Medchemexpress LLC Supplier No. HYW0282631G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Hydroxyflavanone is a naturally occurring flavanone used as a research chemical with reported activity against cyclooxygenase-2 (COX-2), 5-lipoxygenase (5-LOX), and opioid and GABA-A receptors; it is employed in studies of inflammation, neuropathic pain, and diabetes research.

  • High purity (99.87%) suitable for analytical and biological studies.
  • Chemical formula C15H12O3; molecular weight 240.25 g/mol.
  • Available as a powder for convenient storage and handling.
  • Targets COX-2, 5-LOX, opioid receptors, and GABA-A receptors in assays.
  • Reported applications include inflammation, neuropathic pain, and diabetes research.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term).
  • For research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.