Learn More
Medchemexpress LLC 6-Hydroxyflavone | 6665-83-4 | 1 ML
SDP
Catalog No. 5000418290
Encompass_Preferred
6-Hydroxyflavone is an orally effective flavonoid compound. It demonstrates anti-inflammatory effects by inhibiting LPS-induced NO production and promotes osteoblast differentiation by activating AKT, ERK 1/2, and JNK signaling pathways. Additionally, it offers kidney protective effects, inhibits bovine hemoglobin (BHb) glycosylation, and enhances GABA-induced current through specific GABAA receptor subtypes, contributing to an anti-anxiety role. For research use only.
  • Orally effective flavonoid compound.
  • Exhibits anti-inflammatory properties.
  • Promotes osteoblast differentiation.
  • Inhibits bovine hemoglobin (BHb) glycosylation.
  • Offers kidney protective effects.
  • Enhances GABA-induced current via specific GABAA receptors (anti-anxiety).
  • Maintains cell viability at concentrations up to 50 μM.
  • Tested in various cell-based and animal models.
Catalog No. 50-004-18290 Supplier Medchemexpress LLC Supplier No. HYN711010MM/1ML
6-Hydroxyflavone is an orally effective flavonoid compound. It demonstrates anti-inflammatory effects by inhibiting LPS-induced NO production and promotes osteoblast differentiation by activating AKT, ERK 1/2, and JNK signaling pathways. Additionally, it offers kidney protective effects, inhibits bovine hemoglobin (BHb) glycosylation, and enhances GABA-induced current through specific GABAA receptor subtypes, contributing to an anti-anxiety role. For research use only.
  • Orally effective flavonoid compound.
  • Exhibits anti-inflammatory properties.
  • Promotes osteoblast differentiation.
  • Inhibits bovine hemoglobin (BHb) glycosylation.
  • Offers kidney protective effects.
  • Enhances GABA-induced current via specific GABAA receptors (anti-anxiety).
  • Maintains cell viability at concentrations up to 50 μM.
  • Tested in various cell-based and animal models.