Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Hydroxyflavone | 6665-83-4 | 500 MG
SDP

Catalog No. 5000418291
Encompass_Preferred

6-Hydroxyflavone is an orally effective flavonoid compound. It inhibits LPS-induced NO production and has anti-inflammatory effects. It promotes osteoblast differentiation by activating AKT, ERK 1/2, and JNK signaling pathways. 6-Hydroxyflavone also shows an inhibitory effect on bovine hemoglobin (BHb) glycosylation and has a kidney protective effect. Additionally, it enhances GABA-induced current through the Benzodiazepine sites of γ-aminobutyric acid (GABAA) receptors, with a clear preference for α2- and α3- subtypes, playing an anti-anxiety role.

  • Orally effective flavonoid compound
  • Anti-inflammatory effects
  • Promotes osteoblast differentiation
  • Inhibits bovine hemoglobin (BHb) glycosylation
  • Kidney protective effect
  • Enhances GABA-induced current through GABAA receptors, showing anti-anxiety effects
  • Purity: 99.92%
  • Appearance: Solid
  • Color: Light yellow to yellow

Catalog No. 50-004-18291 Supplier Medchemexpress LLC Supplier No. HYN7110500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Hydroxyflavone is an orally effective flavonoid compound. It inhibits LPS-induced NO production and has anti-inflammatory effects. It promotes osteoblast differentiation by activating AKT, ERK 1/2, and JNK signaling pathways. 6-Hydroxyflavone also shows an inhibitory effect on bovine hemoglobin (BHb) glycosylation and has a kidney protective effect. Additionally, it enhances GABA-induced current through the Benzodiazepine sites of γ-aminobutyric acid (GABAA) receptors, with a clear preference for α2- and α3- subtypes, playing an anti-anxiety role.

  • Orally effective flavonoid compound
  • Anti-inflammatory effects
  • Promotes osteoblast differentiation
  • Inhibits bovine hemoglobin (BHb) glycosylation
  • Kidney protective effect
  • Enhances GABA-induced current through GABAA receptors, showing anti-anxiety effects
  • Purity: 99.92%
  • Appearance: Solid
  • Color: Light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.