Learn More
Selleck Chemical LLC 6-Hydroxyflavone (6-HF) S3634-25mg
SDP
Catalog No. 503108827
Encompass_Preferred
6-Hydroxyflavone a naturally occurring flavonoid binds to GABAA receptors with moderate affinity and acts as a partial agonist of GABAA receptors
Catalog No. 50-310-8827 Supplier Selleck Chemical LLC Supplier No. S363425MG
6-Hydroxyflavone a naturally occurring flavonoid binds to GABAA receptors with moderate affinity and acts as a partial agonist of GABAA receptors