Learn More
Medchemexpress LLC 6-Hydroxyindole | 2380-86-1 | 133.15 | 1 ML
SDP
Catalog No. 5000445795
Encompass_Preferred
6-Hydroxyindole is a biochemical reagent and an endogenous long-lasting OATP1B1 inhibitor, often elevated in the plasma of renal failure patients. It is intended strictly for research purposes and not for medical, diagnostic, or food use.
  • Biochemical reagent
  • Endogenous long-lasting OATP1B1 inhibitor
  • Elevated in plasma of renal failure patients
  • For research purposes only
Catalog No. 50-004-45795 Supplier Medchemexpress LLC Supplier No. HYW00243810MM/1ML
6-Hydroxyindole is a biochemical reagent and an endogenous long-lasting OATP1B1 inhibitor, often elevated in the plasma of renal failure patients. It is intended strictly for research purposes and not for medical, diagnostic, or food use.
  • Biochemical reagent
  • Endogenous long-lasting OATP1B1 inhibitor
  • Elevated in plasma of renal failure patients
  • For research purposes only