Learn More
Selleck Chemical LLC 6-Hydroxyindole S3315-25mg
SDP
Supplier:  Selleck Chemical LLC S331525MG
Encompass_Preferred
6-Hydroxyindole (1H-indol-6-ol) is an endogenous potent and long-lasting OATP1B1 inhibitor with IC50 of 10 M
Catalog No. 50-311-0465