Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-hydroxykaempferol 3,6-diglucoside | 142674-16-6 | 99.1% | 626.52 g/mol | C27H30O17 | 10MG
SDP

Catalog No. 5000238015
Encompass_Preferred

6-Hydroxykaempferol 3,6-diglucoside is a flavonoid diglucoside provided as a research reagent with reported antiplatelet aggregatory activity. The compound is characterized analytically and accompanied by safety documentation for laboratory use.

  • High purity (99.1% by HPLC).
  • Chemical formula C27H30O17; molecular weight 626.5 g/mol.
  • Available in small quantities (1 mg to 100 mg) for research applications.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied with analytical documents (COA, LCMS, HNMR) and SDS.
  • Intended for research use only.

Catalog No. 50-002-38015 Supplier Medchemexpress LLC Supplier No. HY12532310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Hydroxykaempferol 3,6-diglucoside is a flavonoid diglucoside provided as a research reagent with reported antiplatelet aggregatory activity. The compound is characterized analytically and accompanied by safety documentation for laboratory use.

  • High purity (99.1% by HPLC).
  • Chemical formula C27H30O17; molecular weight 626.5 g/mol.
  • Available in small quantities (1 mg to 100 mg) for research applications.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Supplied with analytical documents (COA, LCMS, HNMR) and SDS.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.