Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 6-Hydroxynicotinaldehyde, >95%, [106984-91-2], C12919 123.109 C6H5NO2

Catalog No. 501408664
Encompass_Preferred

Compound: 6-Hydroxynicotinaldehyde

CAS#: 106984-91-2

MDL#: not listed

Background: A hydroxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The tautomeric hydroxyl/oxo group allows for further derivatization, while the aldehyde provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-8664 Supplier Frontier Specialty Chemicals Supplier No. C129195G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Hydroxynicotinaldehyde

CAS#: 106984-91-2

MDL#: not listed

Background: A hydroxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The tautomeric hydroxyl/oxo group allows for further derivatization, while the aldehyde provides a reactive handle for reductive amination.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.