Learn More
Medchemexpress LLC 6-Hydroxynicotinic acid | 5006-66-6 | 139.11 | 250 MG
SDP
Catalog No. 5000449969
Encompass_Preferred
6-Hydroxynicotinic acid is an analytical standard for research and analytical applications. It serves as an intermediate in the oxidation of nicotinic acid by Pseudomonas fluorescens and is also a metabolite. This compound can be isolated from root exudates of Cucumis melo, where it disrupts the Auxin signaling pathway and inhibits haustoria precursor formation in Phlipanche aegyptiaca.
  • Analytical standard for research and analytical applications
  • Intermediate in the oxidation of nicotinic acid by Pseudomonas fluorescens and a metabolite
  • Can be isolated from root exudates of Cucumis melo
  • Disrupts Auxin signaling pathway
  • Inhibits haustoria precursor formation in Phlipanche aegyptiaca
  • Commonly used as a reference standard in qualitative, quantitative, and methodological research experiments using HPLC, GC, and MS
Catalog No. 50-004-49969 Supplier Medchemexpress LLC Supplier No. 250MGHYW001996R
6-Hydroxynicotinic acid is an analytical standard for research and analytical applications. It serves as an intermediate in the oxidation of nicotinic acid by Pseudomonas fluorescens and is also a metabolite. This compound can be isolated from root exudates of Cucumis melo, where it disrupts the Auxin signaling pathway and inhibits haustoria precursor formation in Phlipanche aegyptiaca.
  • Analytical standard for research and analytical applications
  • Intermediate in the oxidation of nicotinic acid by Pseudomonas fluorescens and a metabolite
  • Can be isolated from root exudates of Cucumis melo
  • Disrupts Auxin signaling pathway
  • Inhibits haustoria precursor formation in Phlipanche aegyptiaca
  • Commonly used as a reference standard in qualitative, quantitative, and methodological research experiments using HPLC, GC, and MS