Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxynicotinic acid, 98+%

Catalog No. AC156400250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156400250 25 g Plastic bottle
AC156400050 5 g Glass bottle
AC156401000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156400250 Supplier Thermo Scientific Chemicals Supplier No. 156400250
Add to Cart

CAS: 5006-66-6 | C6H5NO3 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-66-6
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006277
InChI Key BLHCMGRVFXRYRN-UHFFFAOYSA-N
Synonym 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
PubChem CID 72924
ChEBI CHEBI:16168
IUPAC Name 6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=CC(=O)NC=C1C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 314°C to 316°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 22, 215
Formula Weight 139.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 6-Hydroxynicotinic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-682-9
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.