Learn More
Medchemexpress LLC 6-Hydroxytropinone | 5932-53-6 | 155.19 | 250 MG
SDP
Catalog No. 5000450028
Encompass_Preferred
6-Hydroxytropinone is an alkaloid product that can be isolated from the roots of Atropa belladonna. This product is for research use only and has not been fully validated for medical applications.
  • Alkaloid product
  • Isolated from the roots of Atropa belladonna
  • Solid appearance
  • Yellow to brown color
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
Catalog No. 50-004-50028 Supplier Medchemexpress LLC Supplier No. 250MGHYW008517
6-Hydroxytropinone is an alkaloid product that can be isolated from the roots of Atropa belladonna. This product is for research use only and has not been fully validated for medical applications.
  • Alkaloid product
  • Isolated from the roots of Atropa belladonna
  • Solid appearance
  • Yellow to brown color
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month