Learn More
OHAUS 6 in. Platform
Catalog No. 01804334 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-334 Vortex Attachment
1 options
Catalog No. 01-804-334 Supplier OHAUS Supplier No. 83041428
Add to Cart
Add to Cart
The 6 in. (152 mm) Platform can be used with the Vortex Genie 2 mixers and the Vortex Genie Pulse.
  • It is required with either the Microtube Foam Insert, Microplate Foam Insert or Two-Tier Microplate Foam Insert.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse