Learn More
Andwin Scientific 6 INCH PLATFORM FOR VORTEX
SDP
Catalog No. NC9735986
Encompass_Preferred
The 6-inch Platform is compatible with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse in conjunction with either the Microtube Foam Insert (Part No. 504-0234-00), Microplate Foam Insert (Part No. 504-0235-00) or Two-Tier Microplate Foam Insert (Part No. SI-0510). This product is made to tightly hold foam inserts.
Catalog No. NC9735986 Supplier Andwin Scientific Supplier No. 146600500
The 6-inch Platform is compatible with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse in conjunction with either the Microtube Foam Insert (Part No. 504-0234-00), Microplate Foam Insert (Part No. 504-0235-00) or Two-Tier Microplate Foam Insert (Part No. SI-0510). This product is made to tightly hold foam inserts.