Learn More
Grainger 6-Inch Work Boot M 10 Black PR
Catalog No. 502766841
Encompass_Preferred
The Grainger 16V675 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is rated for continuous duty applications. The 16V675 operates on 115V power and has a full load amp rating of 12.2A. This electric motor is ideal for driving pumps, fans, compressors, conveyors, and other industrial equipment.
Catalog No. 50-276-6841 Supplier Grainger Supplier No. 16V675
The Grainger 16V675 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is rated for continuous duty applications. The 16V675 operates on 115V power and has a full load amp rating of 12.2A. This electric motor is ideal for driving pumps, fans, compressors, conveyors, and other industrial equipment.